Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED12L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557827
Description
Immunogen sequence: LSVTASTRPR SPVGENADEH YSKDHDVKME IFSPMPGESC ENANTSLGRR MSVNCEKLVK REKPRELIFP SNYDLL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
MED12L, also named as KIAA1635, TNRC11L, TRALP and TRALPUSH, belongs to the Mediator complex subunit 12 family. MED12L may be a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. MED12L is a mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. MED12L is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
Specifications
MED12L | |
Polyclonal | |
Unconjugated | |
MED12L | |
A130035F20; A630079M02; AU044581; KIAA1635; Kiaa3007; MED12L; mediator complex subunit 12 like; mediator complex subunit 12-like; mediator complex subunit 12-like protein; Mediator of RNA polymerase II transcription subunit 12-like protein; mediator of RNA polymerase II transcription, subunit 12 homolog-like; no opposite paired repeat protein; NOPAR; PRO0314; RGD1304617; thyroid hormone receptor-associated-like protein; TNRC11L; TRALP; TRALPUSH; trinucleotide repeat-containing gene 11 protein-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86YW9 | |
MED12L | |
Recombinant protein corresponding to Human MED12L. Recombinant protein control fragment (Product #RP-96365). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction