Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579654
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579654 Supplier Invitrogen™ Supplier No. PA579654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human U2OS whole cell, human MCF-7 whole cell. ICC/IF: U20S cell. Flow: A431 cell.

This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. The product of this gene is proposed to form a sub-complex with MED12, cyclin C, and CDK8 that can negatively regulate transactivation by mediator. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED13
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene MED13
Gene Accession No. Q5SWW4, Q9UHV7
Gene Alias 1110067M05Rik; activator-recruited cofactor 250 kDa component; ARC250; D030023K18; DRIP250; HSPC221; Kiaa0593; Med13; Mediator complex subunit 13; mediator of RNA polymerase II transcription subunit 13; mediator of RNA polymerase II transcription, subunit 13; mediator of RNA polymerase II transcription, subunit 13 homolog; Thrap1; thyroid hormone receptor associated protein 1; thyroid hormone receptor associated protein, 240 kDa subunit; thyroid hormone receptor-associated protein 1; Thyroid hormone receptor-associated protein complex 240 kDa component; thyroid hormone receptor-associated protein complex component TRAP240; thyroid hormone receptor-associated protein, 240 kDa subunit; Trap240; Vitamin D3 receptor-interacting protein complex component DRIP250; Zmediator complex subunit 13
Gene Symbols MED13
Host Species Rabbit
Immunogen E. coli-derived human MED13 recombinant protein (Position: L61-Y240).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 327987, 9969
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.