Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED14 Monoclonal Antibody (483CT11.1.1)
GREENER_CHOICE

Catalog No. PIMA537563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537563 Supplier Invitrogen™ Supplier No. MA537563
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MED14 Monoclonal Antibody for Western Blot

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED14
Applications Western Blot
Classification Monoclonal
Clone 483CT11.1.1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide; pH 7.4
Gene Med14
Gene Accession No. A2ABV5, O60244
Gene Alias 9930001L01Rik; Activator-recruited cofactor 150 kDa component; ARC150; AU041628; Cofactor required for Sp1 transcriptional activation subunit 2; cofactor required for Sp1 transcriptional activation subunit 2 (150 kDa); cofactor required for Sp1 transcriptional activation, subunit 2; cofactor required for Sp1 transcriptional activation, subunit 2 (150kD); CRSP complex subunit 2; CRSP150; CRSP2; CSRP; CXorf4; DRIP150; ENSMUSG00000073278; EXLM1; Gm641; hRGR1; human homolog of yeast RGR1; MED14; Mediator complex subunit 14; mediator of RNA polymerase II transcription subunit 14; MGC104513; ORF1; RGD1560170; RGR1; RGR1 homolog; thyroid hormone receptor-associated protein complex 170 kDa component; thyroid hormone receptor-associated protein complex component TRAP170; Transcriptional coactivator CRSP150; transcriptional co-activator CRSP150; TRAP170; vitamin D receptor-interacting protein complex component DRIP150; vitamin D3 receptor-interacting protein complex 150 kDa component
Gene Symbols Med14
Host Species Mouse
Immunogen purified His-tagged MED14 protein (fragment).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26896, 9282
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.