Learn More
Invitrogen™ MED14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587913
Description
Immunogen sequence: LDQATLDDME KSVNDDMKRI IPWIQQLKFW LGQQRCKQSI KHLPTISSET LQLSNYSTHP IGNLSKNKLF IKLTRLPQYY IVVEMLEVPN KPTQLSYKYY FMSVNAADRE DSPAMALLLQ QFKENIQDLV FRTKTGKQTR TNAKRKLSDD PCPVESKKTK RAGEMCAFNK VLAHFVAMCD T; Positive Samples: HeLa, SW480, Jurkat, Mouse brain, Mouse pancreas; Cellular Location: Nucleus.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
Specifications
MED14 | |
Polyclonal | |
Unconjugated | |
Med14 | |
9930001L01Rik; Activator-recruited cofactor 150 kDa component; ARC150; AU041628; Cofactor required for Sp1 transcriptional activation subunit 2; cofactor required for Sp1 transcriptional activation subunit 2 (150 kDa); cofactor required for Sp1 transcriptional activation, subunit 2; cofactor required for Sp1 transcriptional activation, subunit 2 (150kD); CRSP complex subunit 2; CRSP150; CRSP2; CSRP; CXorf4; DRIP150; ENSMUSG00000073278; EXLM1; Gm641; hRGR1; human homolog of yeast RGR1; MED14; Mediator complex subunit 14; mediator of RNA polymerase II transcription subunit 14; MGC104513; ORF1; RGD1560170; RGR1; RGR1 homolog; thyroid hormone receptor-associated protein complex 170 kDa component; thyroid hormone receptor-associated protein complex component TRAP170; Transcriptional coactivator CRSP150; transcriptional co-activator CRSP150; TRAP170; vitamin D receptor-interacting protein complex component DRIP150; vitamin D3 receptor-interacting protein complex 150 kDa component | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26896, 317343, 9282 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2ABV5, O60244 | |
Med14 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 470-650 of human MED14 (NP_0042202). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.