Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579655
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, mouse liver tissue.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
Specifications
MED14 | |
Polyclonal | |
Unconjugated | |
Med14 | |
9930001L01Rik; Activator-recruited cofactor 150 kDa component; ARC150; AU041628; Cofactor required for Sp1 transcriptional activation subunit 2; cofactor required for Sp1 transcriptional activation subunit 2 (150 kDa); cofactor required for Sp1 transcriptional activation, subunit 2; cofactor required for Sp1 transcriptional activation, subunit 2 (150kD); CRSP complex subunit 2; CRSP150; CRSP2; CSRP; CXorf4; DRIP150; ENSMUSG00000073278; EXLM1; Gm641; hRGR1; human homolog of yeast RGR1; MED14; Mediator complex subunit 14; mediator of RNA polymerase II transcription subunit 14; MGC104513; ORF1; RGD1560170; RGR1; RGR1 homolog; thyroid hormone receptor-associated protein complex 170 kDa component; thyroid hormone receptor-associated protein complex component TRAP170; Transcriptional coactivator CRSP150; transcriptional co-activator CRSP150; TRAP170; vitamin D receptor-interacting protein complex component DRIP150; vitamin D3 receptor-interacting protein complex 150 kDa component | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26896, 317343, 9282 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
500 μg/mL | |
PBS with 4 mg trehalose and 0.05 mg sodium azide | |
A2ABV5, O60244 | |
Med14 | |
E. coli-derived human MED14 recombinant protein (Position: D181-D375). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction