Learn More
MED15 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302422AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors. Required for cholesterol-dependent gene regulation. Positively regulates the Nodal signaling pathway.Specifications
MED15 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96RN5 | |
MED15 | |
Between 450 and 500 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
MED15 | |
MED15; mediator complex subunit 15; ARC105; CAG7A; CTG7A; DKFZp686A2214; DKFZp762B1216; FLJ42282; FLJ42935; PCQAP; TIG-1; TIG1; TNRC7; CTG repeat protein 7a; PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associated protein; PC2-glutamine-rich-associated protein; TPA inducible gene-1; TPA inducible protein; activator-recruited cofactor, 105-kD; positive cofactor 2, glutamine/Q-rich-associated protein; trinucleotide repeat containing 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51586 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.