Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540839
Description
Peptide sequence: AAQILLKGAE RLTKSVTENQ ENKLQRDFNS ELLRLRQHWK LRKVGDKILG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by CRSP6 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e. g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
Specifications
MED17 | |
Polyclonal | |
Unconjugated | |
MED17 | |
77kDa; activator-recruited cofactor 77 kDa component; ARC77; C330002H14Rik; cofactor required for Sp1 transcriptional activation subunit 6; cofactor required for Sp1 transcriptional activation, subunit 6; cofactor required for Sp1 transcriptional activation, subunit 6 (77kD); cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa; CRSP complex subunit 6; CRSP6; CRSP77; DRIP77; DRIP80; Med17; Mediator complex subunit 17; mediator of RNA polymerase II transcription subunit 17; Thyroid hormone receptor-associated protein complex 80 kDa component; transcriptional coactivator CRSP77; Trap80; Vitamin D3 receptor-interacting protein complex 80 kDa component | |
Rabbit | |
Affinity chromatography | |
RUO | |
9440 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NVC6 | |
MED17 | |
Synthetic peptide directed towards the N-terminal of human CRSP6 (aa 156-205). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction