Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115563 Supplier Invitrogen™ Supplier No. PA5115563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MED17.

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by CRSP6 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e. g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED17
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MED17
Gene Accession No. Q8VCD5, Q9NVC6
Gene Alias 77kDa; activator-recruited cofactor 77 kDa component; ARC77; C330002H14Rik; cofactor required for Sp1 transcriptional activation subunit 6; cofactor required for Sp1 transcriptional activation, subunit 6; cofactor required for Sp1 transcriptional activation, subunit 6 (77kD); cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa; CRSP complex subunit 6; CRSP6; CRSP77; DRIP77; DRIP80; Med17; Mediator complex subunit 17; mediator of RNA polymerase II transcription subunit 17; Thyroid hormone receptor-associated protein complex 80 kDa component; transcriptional coactivator CRSP77; Trap80; Vitamin D3 receptor-interacting protein complex 80 kDa component
Gene Symbols MED17
Host Species Rabbit
Immunogen A synthesized peptide derived from human MED17(Accession Q9NVC6), corresponding to amino acid residues E186-I236.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234959, 9440
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.