Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED17 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | MED17 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
MED17 Polyclonal specifically detects MED17 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MED17 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Activator-recruited cofactor 77 kDa component, Cofactor required for Sp1 transcriptional activation subunit 6, cofactor required for Sp1 transcriptional activation, subunit 6, 77kDa, CRSP6FLJ10812, CRSP77cofactor required for Sp1 transcriptional activation, subunit 6 (77kD), DRIP77, DRIP80ARC77, mediator complex subunit 17Vitamin D3 receptor-interacting protein complex 80 kDa component, mediator of RNA polymerase II transcription subunit 17, Thyroid hormone receptor-associated protein complex 80 kDa component, Transcriptional coactivator CRSP77, Trap80, TRAP80CRSP complex subunit 6 | |
| The immunogen is a synthetic peptide directed towards the N terminal region of human MED17 (NP_004259). Peptide sequence AAQILLKGAERLTKSVTENQENKLQRDFNSELLRLRQHWKLRKVGDKILG | |
| Affinity purified |
| Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS buffer, 2% sucrose | |
| 9440 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title