Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED18 Polyclonal Antibody

MED18 Polyclonal Antibody
Supplier: Thermo Scientific PA555613
Description
MED18 Polyclonal Antibody for ICC/IF, IHC
Specifications
| MED18 | |
| Polyclonal | |
| Unconjugated | |
| MED18 | |
| 2810046C01Rik; FLJ20045; Med18; mediator 18; Mediator complex subunit 18; mediator of RNA polymerase II transcription subunit 18; mediator of RNA polymerase II transcription, subunit 18 homolog; p28b; TRAP/mediator complex subunit p28b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54797 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BUE0 | |
| MED18 | |
| Recombinant protein corresponding to Human MED18. Recombinant protein control fragment (Product #RP-94185). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction