Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120283
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120283 Supplier Invitrogen™ Supplier No. PA5120283
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, Jurkat, Mouse liver, Mouse thymus. The target is usually found in the following locations: Nucleus. Immunogen sequence: MEAPPVTMMP VTGGTINMME YLLQGSVLDH SLESLIHRLR GLCDNMEPET FLDHEMVFLL KGQQASPFVL RARRSMDRAG APWHLRYLGQ PEMGDKNRHA LVRNCVDIAT SENLTDFLME MGFRMDHEFV AKGHLFRKGI MKIMVYKIFR ILVPGNTDST EALSLSYLVE LSVVAPAGQD MVSDDMKNFA EQLKPLVHLE KIDPKRLM.

Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED18
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MED18
Gene Accession No. Q9BUE0, Q9CZ82
Gene Alias 2810046C01Rik; FLJ20045; Med18; mediator 18; Mediator complex subunit 18; mediator of RNA polymerase II transcription subunit 18; mediator of RNA polymerase II transcription, subunit 18 homolog; p28b; TRAP/mediator complex subunit p28b
Gene Symbols MED18
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human MED18 (NP_001120822.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54797, 67219
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.