Learn More
MED18 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | MED18 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1632
![]() |
Bethyl Laboratories, Inc
IHC00181T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1631
![]() |
Bethyl Laboratories, Inc
IHC00181 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monke
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II (Sato et al., 2003 [PubMed 12584197]).Specifications
MED18 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
p28b, TRAP/mediator complex subunit p28b, mediator of RNA polymerase II transcription subunit 18, mediator of RNA polymerase II transcription, subunit 18 homolog | |
Between 150 and C-term | |
Primary | |
4° C | |
MED18 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9BUE0, Q9CZ82 | |
54797, 67219 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.