Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559329 Supplier Invitrogen™ Supplier No. PA559329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPGADKSGAG CGPFYLMREL PGSTELTGST NLITHYNLEQ AYNKFCGKKV KEKLSNFLPD LPGMIDLPG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

MED19 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED19
Gene Accession No. A0JLT2
Gene Alias 2410018M14Rik; 3110040A13Rik; DT2P1G7; LCMR1; Lung cancer metastasis-related protein 1; Med19; MED19AS; mediator complex subunit 19; Mediator of RNA polymerase II transcription subunit 19; mediator of RNA polymerase II transcription, subunit 19 homolog; putative mediator subunit MED19AS protein; RGD1311926
Gene Symbols MED19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MED19. Recombinant protein control fragment (Product #RP-97401).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.