Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED20 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP184140
Description
MED20 Polyclonal specifically detects MED20 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MED20 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
hTRFP, mediator complex subunit 20MGC29869, mediator of RNA polymerase II transcription subunit 20, PRO0213, Trf (TATA binding protein-related factor)-proximal homolog, Trf (TATA binding protein-related factor)-proximal homolog (Drosophila), TRFPDKFZp586D2223, TRF-proximal protein homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MED20 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EKQGTFCVDCETYHTAASTLGSQGQTGKLMYVMHNSEYPLSCFALFENGPCLIADTNFDVLMVKLKGFFQSAKASKIETRGTRYQYCDFLVKVGTVTMGPSARGISVEVEYGPCVV | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
9477 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction