Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100799
Description
Antibody detects endogenous levels of total MED20.
MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
Specifications
MED20 | |
Polyclonal | |
Unconjugated | |
MED20 | |
1110011O05Rik; 2410115I17Rik; AU018348; hTRFP; Med20; Mediator complex subunit 20; mediator of RNA polymerase II transcription subunit 20; PRO0213; Trf (TATA binding protein-related factor)-proximal homolog; Trf (TATA binding protein-related factor)-proximal protein homolog; TRFP; TRF-proximal protein homolog; ubiquitin specific protease 49; Usp49 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
316209, 56771, 9477 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5XIE9, Q9H944, Q9R0X0 | |
MED20 | |
A synthesized peptide derived from human MED20(Accession Q9H944), corresponding to amino acid residues K27-A77. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction