Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588393 Supplier Invitrogen™ Supplier No. PA588393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADRLTQLQD AVNSLADQFC NAIGVLQQCG PPASFNNIQT AINKDQPANP TEEYAQLFAA LIARTAKDID VLIDSLPSEE STAALQAASL YKLEEENHEA ATCLEDVVYR GDMLLEKIQS ALADIAQSQL KTRSGTHSQS LPDS; Positive Samples: LO2, U-87MG, 293T, A-549, 22RV1, Mouse liver, Mouse testis; Cellular Location: Nucleus.

This gene encodes a member of the mediator complex subunit 21 family. The encoded protein interacts with the human RNA polymerase II holoenzyme and is involved in transcriptional regulation of RNA polymerase II transcribed genes. A pseudogene of this gene is located on chromosome 8. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED21
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MED21
Gene Accession No. Q13503, Q9CQ39
Gene Alias 0610007L03Rik; AI449604; D19234; D6Ertd782e; fk48d02; hSrb7; hypothetical protein LOC539496; MED21; Mediator complex subunit 21; mediator of RNA polymerase II transcription subunit 21; RNA polymerase II holoenzyme component SRB7; RNAPII complex component SRB7; si:ch211-203h15.4; SRB7; SRB7 (supressor of RNA polymerase B) homolog; SRB7 suppressor of RNA polymerase B homolog; surb7; wu:fk48d02; zgc:56532
Gene Symbols MED21
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-144 of human MED21 (NP_0042552).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108098, 9412
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.