Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589465 Supplier Invitrogen™ Supplier No. PA589465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQQRALPQS KETLLQSYNK RLKDDIKSIM DNFTEIIKTA KIEDETQVSR ATQGEQDNYE MHVRAANIVR AGESLMKLVS DLKQFLILND FPSVNEAIDQ RNQQLRTLQE ECDRKLITLR DEISIDLYEL EEEYYSSRYK; Positive Samples: U-87MG, A-431, A-549, Mouse testis; Cellular Location: Nucleus.

This gene is located in the surfeit gene cluster, a group of very tightly linked housekeeping genes that do not share sequence similarity. The gene is oriented in a head-to-head fashion with RPL7A (SURF3) and the two genes share a bidirectional promoter. The encoded proteins are localized to the cytoplasm. Two alternative transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED22
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MED22
Gene Accession No. A0JPN6, Q15528, Q62276
Gene Alias AW212655; AW558812; MED22; MED24; Mediator complex subunit 22; mediator of RNA polymerase II transcription subunit 22; MGC48682; RGD1564893; si:ch73-278c20.1; si:dkey-226m8.2; surf5; Surf-5; surfeit 5; surfeit 5 isoform b; surfeit gene 5; surfeit locus protein 5; wu:fj45h07; zgc:86721
Gene Symbols MED22
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human MED22 (NP_8524681).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20933, 499762, 6837
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.