Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED23 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB200339AF532
Description
MED23 Polyclonal antibody specifically detects MED23 in Human samples. It is validated for Western BlotSpecifications
MED23 | |
Polyclonal | |
Western Blot | |
130 kDa transcriptional co-activator, 133 kDa transcriptional co-activator, CRSP130, DRIP130DKFZp434H0117, mediator complex subunit 23CRSP133, Protein sur-2 homolog, subunit 3, 130kDa, Transcriptional coactivator CRSP130, Vitamin D3 receptor-interacting protein complex 130 kDa component | |
between residues 1300 and the C-terminus of human MED23 protein | |
0.1 mL | |
Cell Biology, Signal Transduction, Transcription Factors and Regulators | |
9439 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction