Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074599
Description
MED24 Polyclonal specifically detects MED24 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MED24 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa, CRSP100, DRIP100CRSP complex subunit 4, hTRAP100, KIAA0130CRSP4, mediator complex subunit 24ARC100, MGC8748, subunit 24 homolog, thyroid hormone receptor associated protein 4, Thyroid hormone receptor-associated protein 4, Thyroid hormone receptor-associated protein complex 100 kDa component, Trap100, TRAP100THRAP4, Vitamin D3 receptor-interacting protein complex 100 kDa component, vitamin D3 receptor-interacting protein complex component DRIP100 | |
Rabbit | |
110 kDa | |
100 μL | |
Primary | |
MED24. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
O75448 | |
MED24 | |
The immunogen recognized by this antibody maps to a region between residue 939 and 989 of human thyroid hormone receptor-associated protein complex 100 kDa component (mediator complex subunit 24 ) using the numbering given in entry NP_055630.2 (GeneID 986). | |
Affinity Purified | |
RUO | |
9862 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction