Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MED24 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MED24 Polyclonal specifically detects MED24 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MED24 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
9862 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa, CRSP100, DRIP100CRSP complex subunit 4, hTRAP100, KIAA0130CRSP4, mediator complex subunit 24ARC100, MGC8748, subunit 24 homolog, thyroid hormone receptor associated protein 4, Thyroid hormone receptor-associated protein 4, Thyroid hormone receptor-associated protein complex 100 kDa component, Trap100, TRAP100THRAP4, Vitamin D3 receptor-interacting protein complex 100 kDa component, vitamin D3 receptor-interacting protein complex component DRIP100 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human MED24 (NP_055630.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title