Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584792 Supplier Invitrogen™ Supplier No. PA584792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPGAASGPPP PGPILRPQNP GANPQLRSLL LNPPPPQTGV PPPQASLHHL QPPG Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes a component of the transcriptional coactivator complex termed the Mediator complex. This complex is required for transcription of most RNA polymerase II-dependent genes. The encoded protein plays a role in chromatin modification and in preinitiation complex assembly. Mutations in this gene are associated with Charcot-Marie-Tooth disease type 2B2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED25
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED25
Gene Accession No. Q71SY5
Gene Alias 2610034E13Rik; 2610529E18Rik; ACID1; Activator interaction domain-containing protein 1; activator-recruited cofactor 92 kDa component; ARC/mediator transcriptional coactivator subunit; ARC92; BVSYS; CMT2B2; DKFZp434K0512; ESTM2; hypothetical protein LOC406793; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 25; MED25; Mediator complex subunit 25; mediator of RNA polymerase II transcription subunit 25; mediator of RNA polymerase II transcription, subunit 25 homolog; mediator of RNA polymerase II transcription, subunit 9 homolog; MGC70671; mMED25; P78; prostate-derived protein 78 kDa; PTOV2; RGD1305137; TCBAP0758; zgc:63555
Gene Symbols MED25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MED25. Recombinant protein control fragment (Product #RP-105458).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.