Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Med26 Polyclonal Antibody, Bioss
SDP

Catalog No. 502281960
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1960 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1960 Supplier Bioss Supplier No. BS53124R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Med26 Polyclonal antibody specifically detects Med26 in Human samples. It is validated for Western Blot

Specifications

Antigen Med26
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene MED26
Gene Accession No. O95402
Gene Alias 5730493L18Rik; Activator-recruited cofactor 70 kDa component; AI414941; ARC70; AW495270; cofactor required for Sp1 transcriptional activation subunit 7; cofactor required for Sp1 transcriptional activation, subunit 7 (70kD); cofactor required for Sp1 transcriptional activation, subunit 7, 70kDa; CRSP complex subunit 7; Crsp7; CRSP70; Med26; Mediator complex subunit 26; mediator of RNA polymerase II transcription subunit 26; Slc35e1; solute carrier family 35, member E1; transcriptional coactivator CRSP70
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against human Med26, using a KLH-conjugated synthetic peptide containing a sequence from the N-terminal part of the protein.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9441
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.