Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED28 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297494MFV500
Description
MED28 Polyclonal antibody specifically detects MED28 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
MED28 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
DKFZP434N185, EG1DKFZp434N185, magicin, mediator complex subunit 281500003D12Rik, mediator of RNA polymerase II transcription, subunit 28 homolog (S. cerevisiae), subunit 28 homolog | |
Produced in rabbits immunized with E. coli-derived Human MED28 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
80306 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction