Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED28 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557456 Supplier Invitrogen™ Supplier No. PA557456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSSTLVDEL ESSFEACFAS LVSQDYVNGT DQEEIRTGVD QCIQKFLDIA RQTECFFLQK RLQLSVQKPE QV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The mediator complex is a multi-protein transcriptional co-activator that is expressed ubiquitously in eukaryotes from yeast to mammals and is required for induction of RNA polymerase II (pol II) transcription by DNA binding transcription factor. One of the proteins in this complex is MED28, also known as Magicin. MED28 is expressed in many cell lines and tissues. It has been shown that a down-regulation of MED28 expression in NIH3T3 cells results in a significant induction of several genes associated with smooth muscle cell differentiation and conversely its over-expression represses expression of SMC genes. MED28 can also form a ternary complex with Grb2 and Merlin, the neurofibromatosis 2 tumor suppressor protein, indicating that MED28 may play a role in Merlin's tumor suppressive activity. MED28 has also been recently identified as an HIV dependency factor (HDF), suggesting that MED28 may be an important drug target in HIV treatment. At least two isoforms of MED28 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED28
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED28
Gene Accession No. Q9H204
Gene Alias 1500003D12Rik; AI451633; AU045690; EG1; endothelial-derived; endothelial-derived gene 1; endothelial-derived protein 1; Fksg20; Magicin; Med28; Mediator complex subunit 28; mediator of RNA polymerase II transcription subunit 28; mediator of RNA polymerase II transcription, subunit 28 homolog; merlin and Grb2-interacting cytoskeletal protein; RGD1305875; Tumor angiogenesis marker EG-1
Gene Symbols MED28
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MED28. Recombinant protein control fragment (Product #RP-102687).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80306
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.