Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED29 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575625 Supplier Invitrogen™ Supplier No. PA575625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multi-protein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med29 (mediator complex subunit 29), also known as IXL (intersex-like), is a 200 amino acid nuclear protein and component of the mediator complex. Widely expressed in embryo and adult tissue, Med29 is considered a novel amplification target gene in pancreatic cancer and is encoded by a gene that maps to human chromosome 19q13.2. Chromosome 19 consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED29
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MED29
Gene Accession No. Q9DB91, Q9NX70
Gene Alias 2810405O22Rik; AU020902; chunp6869; DKFZp434H247-like protein; intersex-like protein; ixl; ixl protein; MED2; MED29; Mediator complex subunit 29; mediator of RNA polymerase II transcription subunit 29; RGD1311009; similar to Homo sapiens DKFZp434H247 protein; zgc:109789
Gene Symbols MED29
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human MED29.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292751, 55588, 67224
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.