Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MED29 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568220
Description
Predicted to react with Mouse samples.
In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multi-protein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med29 (mediator complex subunit 29), also known as IXL (intersex-like), is a 200 amino acid nuclear protein and component of the mediator complex. Widely expressed in embryo and adult tissue, Med29 is considered a novel amplification target gene in pancreatic cancer and is encoded by a gene that maps to human chromosome 19q13.2. Chromosome 19 consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.
Specifications
MED29 | |
Polyclonal | |
Unconjugated | |
MED29 | |
2810405O22Rik; AU020902; chunp6869; DKFZp434H247-like protein; intersex-like protein; ixl; ixl protein; MED2; MED29; Mediator complex subunit 29; mediator of RNA polymerase II transcription subunit 29; RGD1311009; similar to Homo sapiens DKFZp434H247 protein; zgc:109789 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55588, 67224 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9DB91, Q9NX70 | |
MED29 | |
Synthesized peptide derived from human MED29. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction