Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED30 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287793
Description
MED30 Polyclonal specifically detects MED30 in Human samples. It is validated for Western Blot.Specifications
MED30 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
mediator complex subunit 30TRAP/Mediator complex component TRAP25, mediator of RNA polymerase II transcription subunit 30, putative mediator of RNA polymerase II transcription subunit 30, THRAP6MGC9890, thyroid hormone receptor associated protein 6, Thyroid hormone receptor-associated protein 6, Thyroid hormone receptor-associated protein complex 25 kDa component, Trap25, TRAP25MED30S | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human MED30 | |
100 μL | |
Signal Transduction | |
90390 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction