Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565417 Supplier Invitrogen™ Supplier No. PA565417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSTPPLAASGM APGPFAGPQA QQAAREVNTA SLCRIGQETV QDIVYRTMEI FQLLRNMQLP NGVTY Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

Med30 is a subunit of the RNA polymerase II (Pol II) transcriptional mediator complex. The mediator complex is a coactivator involved in the regulated transcription of Pol II-dependent genes. The mediator complex functions as a bridge to convey information from gene-specific regulatory proteins to the basal Pol II transcription machinery, and is recruited to promoter regions by directly interacting with regulatory proteins. The mediator complex also serves as a scaffold for the assembly of a functional pre-initiation complex with Pol II and other general transcription factors. Found in the nucleus, Med30 is expressed in brain, heart, kidney, liver, lung, pancreas, placenta and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED30
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED30
Gene Accession No. Q96HR3
Gene Alias 1810038N03Rik; 2510044J04Rik; Med30; MED30S; Mediator complex subunit 30; mediator of RNA polymerase II transcription subunit 30; putative mediator of RNA polymerase II transcription subunit 30; THRAP6; thyroid hormone receptor associated protein 6; thyroid hormone receptor-associated protein 6; Thyroid hormone receptor-associated protein complex 25 kDa component; TRAP/Mediator complex component TRAP25; TRAP25
Gene Symbols MED30
Host Species Rabbit
Immunogen Recombinant Human MED30. Recombinant protein control fragment (Product #RP-106097).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90390
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.