Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED30 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549159
Description
Predicted to react with mouse based on sequence homology.
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
Specifications
MED30 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96HR3 | |
MED30 | |
KLH conjugated synthetic peptide between 107-133 amino acids from the Central region of human MED30. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MED30 | |
Mediator of RNA polymerase II transcription subunit 30, Mediator complex subunit 30, TRAP/Mediator complex component TRAP25, Thyroid hormone receptor-associated protein 6, Thyroid hormone receptor-associated protein complex 25 kDa component, Trap25, MED30, THRAP6, TRAP25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90390 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction