Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570247 Supplier Invitrogen™ Supplier No. PA570247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.

MED31 is the component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED31
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MED31
Gene Accession No. 0
Gene Alias 3110004H13Rik; CGI-125; fa04d01; FLJ27436; FLJ36714; hSOH1; l11Jus15; med31; Mediator complex subunit 31; mediator complex subunit soh1; Mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription subunit 31; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription, subunit 31 homolog; RGD1309457; soh1; wu:fa04d01; zgc:92673
Gene Symbols MED31
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of rat MED31.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287475
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.