Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED37A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599016 Supplier Invitrogen™ Supplier No. PA599016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED37A Polyclonal Antibody for Western Blot, ELISA

In cooperation with other chaperones, Hsp70s are key components that facilitate folding of de novo synthesized proteins, assist translocation of precursor proteins into organelles, and are responsible for degradation of damaged protein under stress conditions (Probable). Involved in polar nuclei fusion during female gametophyte development and is essential for the regulation of endosperm nuclei proliferation (PubMed:20080634). Involved in sperm nuclear fusion with central cell polar nuclei at fertilization, which is critical for normal endosperm nuclear proliferation (PubMed:31410484). Required for pollen development and pollen tube growth (PubMed:24486762). Possesses ATPase activity in vitro (PubMed:26186593). [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED37A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Alias AtBP11; AtHsp70-111; BIP1; Heat shock 70 kDa protein 111; Heat shock protein 70-111; HSP70-111; Luminal-binding protein 1; MED37_5
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana Mediator of RNA polymerase II transcription subunit 37a protein (28-669AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.