Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520571
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520571 Supplier Invitrogen™ Supplier No. PA520571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human testis tissue lysate. PA5-20571 can be used with blocking peptide PEP-0691.

The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (Sato et al., 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MED4
Gene Accession No. Q561Q8, Q9CQA5, Q9NPJ6
Gene Alias 2410046H15Rik; Activator-recruited cofactor 36 kDa component; ARC36; DRIP36; FLJ10956; HSPC126; hypothetical protein LOC550582; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 4; MED4; Mediator complex subunit 4; mediator of RNA polymerase II transcription subunit 4; mediator of RNA polymerase II transcription, subunit 4 homolog; mediator, 34-kD subunit, homolog; p36 TRAP/SMCC/PC2 subunit; RP11-90M2.2; TRAP/SMCC/PC2 subunit p36; TRAP/SMCC/PC2 subunit p36 subunit; TRAP36; Vdirp; Vdrip; vitamin D receptor interacting protein; vitamin D receptor-interacting protein, 36-kD; vitamin D3 receptor-interacting protein complex 36 kDa component; zgc:110646
Gene Symbols MED4
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the carboxy terminus of human MED4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29079, 306030, 67381
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.