Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581522
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581522 Supplier Invitrogen™ Supplier No. PA581522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MED4.

The Mediator is a multiprotein coactivator that is required by DNA-binding transcription factors for activation of polymerase II transcribed genes. MED4 appears to be a core Mediator subunit and is found in nearly all Mediator preparations (Sato et al., 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MED4
Gene Accession No. Q9NPJ6
Gene Alias 2410046H15Rik; Activator-recruited cofactor 36 kDa component; ARC36; DRIP36; FLJ10956; HSPC126; hypothetical protein LOC550582; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 4; MED4; Mediator complex subunit 4; mediator of RNA polymerase II transcription subunit 4; mediator of RNA polymerase II transcription, subunit 4 homolog; mediator, 34-kD subunit, homolog; p36 TRAP/SMCC/PC2 subunit; RP11-90M2.2; TRAP/SMCC/PC2 subunit p36; TRAP/SMCC/PC2 subunit p36 subunit; TRAP36; Vdirp; Vdrip; vitamin D receptor interacting protein; vitamin D receptor-interacting protein, 36-kD; vitamin D3 receptor-interacting protein complex 36 kDa component; zgc:110646
Gene Symbols MED4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human MED4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29079
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.