Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED6 Antibody (OTI3C9), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272620IR
Description
MED6 Monoclonal specifically detects MED6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MED6 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MED6 | |
Full length recombinant protein of human MED6 (NP_005457) produced in E.coli. | |
0.1 mL | |
10001.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3C9 | |
Western Blot | |
Activator-recruited cofactor 33 kDa component, ARC33, mediator complex subunit 6hMed6, mediator of RNA polymerase II transcription subunit 6, mediator of RNA polymerase II transcription, subunit 6 homolog, mediator of RNA polymerase II transcription, subunit 6 homolog (S. cerevisiae), NY-REN-28, Renal carcinoma antigen NY-REN-28 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction