Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED7 Antibody (PCRP-MED7-1B8), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314071V
This item is not returnable.
View return policy
Description
MED7 Monoclonal antibody specifically detects MED7 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| MED7 | |
| Monoclonal | |
| DyLight 405 | |
| Activator-recruited cofactor 34 kDa component, Cofactor required for Sp1 transcriptional activation subunit 9, cofactor required for Sp1 transcriptional activation, subunit 9 (33kD), cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa, CRSP33CRSP complex subunit 9, CRSP9hMED7, mediator complex subunit 7ARC34, mediator of RNA polymerase II transcription subunit 7, MGC12284, RNA polymerase transcriptional regulation mediator subunit 7 homolog, Transcriptional coactivator CRSP33 | |
| Recombinant full-length human MED7 protein (Uniprot: O43513) | |
| 0.1 mL | |
| DNA Replication Transcription Translation and Splicing | |
| 9443 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-MED7-1B8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction