Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED7 Mouse anti-Human, Clone: PCRP-MED7-1B8, Novus Biologicals™
SDP

Catalog No. NB177589 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177589 20 μg
NB177588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177589 Supplier Novus Biologicals Supplier No. NBP31381620UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MED7 Monoclonal antibody specifically detects MED7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein Array

Specifications

Antigen MED7
Applications Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array
Classification Monoclonal
Clone PCRP-MED7-1B8
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. O43513
Gene Alias Activator-recruited cofactor 34 kDa component, Cofactor required for Sp1 transcriptional activation subunit 9, cofactor required for Sp1 transcriptional activation, subunit 9 (33kD), cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa, CRSP33CRSP complex subunit 9, CRSP9hMED7, mediator complex subunit 7ARC34, mediator of RNA polymerase II transcription subunit 7, MGC12284, RNA polymerase transcriptional regulation mediator subunit 7 homolog, Transcriptional coactivator CRSP33
Host Species Mouse
Immunogen Recombinant full-length human MED7 protein (Uniprot: O43513)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline DNA Replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9443
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.