Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579661
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579661 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579661 Supplier Invitrogen™ Supplier No. PA579661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, rat heart tissue, rat kidney tissue, mouse brain tissue, mouse heart tissue, mouse kidney tissue. IHC: human colon cancer tissue, human lung cancer tissue, mouse small intestine tissue, rat small intestine tissue, human mammary cancer tissue. ICC/IF: U20S cell. Flow: MCF-7 cell.

In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multi-protein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med9 (mediator complex subunit 9), also known as MED25, is a 146 amino acid nuclear protein and component of the mediator complex. Med9 directly interacts with Med4 and is encoded by a gene that maps to human chromosome 17p11.2. Chromosome 17 comprises over 2.5% of the human genome, encodes over 1,200 genes and is associated with two key tumor suppressor genes, namely, p53 and BRCA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED9
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Med9
Gene Accession No. Q8VCS6, Q9NWA0
Gene Alias BC019367; Med25; Med9; Mediator complex subunit 9; mediator of RNA polymerase II transcription subunit 9; mediator of RNA polymerase II transcription, subunit 9 homolog; mediator subunit 25; RGD1563669
Gene Symbols Med9
Host Species Rabbit
Immunogen E. coli-derived human MED9 recombinant protein (Position: A55-E146).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192191, 497914, 55090
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.