Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEDAG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570956 Supplier Invitrogen™ Supplier No. PA570956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 93%.

MEDAG is a protein coding gene involved in processes that promote adipocyte differentiation, lipid accumulation, and glucose uptake in mature adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEDAG
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MEDAG
Gene Accession No. Q5VYS4
Gene Alias 6330406I15Rik; Activated in W/Wv mouse stomach 3; activated in W/Wv mouse stomach 3 homolog; Awms3; C12H13orf33; C13orf33; C25H13orf33; hAWMS3; mAWMS3; MEDA4; MEDA-4; Medag; mesenteric estrogen dependent adipogenesis; mesenteric estrogen-dependent adipogenesis; mesenteric estrogen-dependent adipogenesis protein; Mesenteric estrogen-dependent adipose 4; RGD1307396; uncharacterized protein C13orf33 homolog
Gene Symbols MEDAG
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MEDAG.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84935
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.