Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEDAG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567634
Description
Predicted to react with Mouse samples.
MEDAG is a protein coding gene involved in processes that promote adipocyte differentiation, lipid accumulation, and glucose uptake in mature adipocytes.
Specifications
MEDAG | |
Polyclonal | |
Unconjugated | |
MEDAG | |
6330406I15Rik; Activated in W/Wv mouse stomach 3; activated in W/Wv mouse stomach 3 homolog; Awms3; C12H13orf33; C13orf33; C25H13orf33; hAWMS3; mAWMS3; MEDA4; MEDA-4; Medag; mesenteric estrogen dependent adipogenesis; mesenteric estrogen-dependent adipogenesis; mesenteric estrogen-dependent adipogenesis protein; Mesenteric estrogen-dependent adipose 4; RGD1307396; uncharacterized protein C13orf33 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
70717, 84935 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q14BA6, Q5VYS4 | |
MEDAG | |
Synthesized peptide derived from human C13orf33. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction