Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Aspirin Tablets

Catalog No. 19090790 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box
2/Pk., 50/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-790 2/Pk., 250/Box
19-090-792 2/Pk., 50/Box
19-090-791 2/Pk., 125/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-790 Supplier Medique Supplier No. 80513
Add to Cart
Add to Cart

For the relief of fever, headache, minor muscular aches, arthritis pain.

  • Easy-to-swallow film-coated tablets
  • In unit dose packets
  • With Spanish labeling where possible

Specifications

Product Type Aspirin
For Use With (Application) Pain, ache, and fever relief
Format Film-coated Tablet
Quantity 2/Pk., 250/Box
Recommended Age 12 years and olde
Storage Requirements 59°F to 86°F (15°C to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.