Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Aspirin Tablets

Catalog No. p-7078216
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box
2/Pk., 50/Box

For the relief of fever, headache, minor muscular aches, arthritis pain.

  • Easy-to-swallow film-coated tablets
  • In unit dose packets
  • With Spanish labeling where possible

Specifications

For Use With (Application) Pain, ache, and fever relief
Product Type Aspirin
Format Film-coated Tablet
Quantity 2/Pk., 50/Box
Recommended Age 12 years and olde
Storage Requirements 59°F to 86°F (15°C to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.