Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Cough Drops

Catalog No. 19090809 Shop All Medique Products
Change view
Click to view available options
Quantity:
1/Pk., 125/Box
1/Pk., 50/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-809 1/Pk., 125/Box
19-090-819 1/Pk., 50/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-090-809 Supplier Medique Supplier No. 81525
Add to Cart
Add to Cart

With vapor action for temporary relief of cough and dry throat.

  • Cough suppressant
  • Each lozenge contains Menthol 7.6mg and eucalyptus

Specifications

For Use With (Application) For temporary relief of coughing and dry throat
Active Ingredients Menthol (7.6 mg), Eucalyptus
Product Type Cough Drops
Flavor Cherry
Quantity 1/Pk., 125/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.