Learn More
Medique Medi-First™ Extra-Strength Non-Aspirin Tablets
Catalog No. 19090800 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box
2/Pk., 50/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-800 2/Pk., 125/Box
19-090-801 2/Pk., 50/Box
19-090-799 2/Pk., 250/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-090-800 Supplier Medique Supplier No. 80448
Add to Cart
Add to Cart
For pain and fever relief.
  • High potency
  • Each tablet contains Acetaminophen 500mg
Specifications
Product Type Extra Strength Non-aspirin Tablets
Active Ingredients Acetaminophen (500 mg)
Quantity 2/Pk., 125/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)