Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Sinus Decongestant

Catalog No. p-7078223
Click to view available options
Quantity:
1/Pk., 100/Box
1/Pk., 250/Box
1/Pk., 500/Box

Clears sinus pressure and nasal congestion.

  • Non-drowsy formula
  • Each tablet contains Phenylephrine HCl 10mg

Specifications

Active Ingredients Phenylephrine HCl (10 mg)
For Use With (Application) Clears sinus pressure and nasal congestion
Product Type Non-drowsy Sinus Decongestant
Quantity 1/Pk., 100/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.