Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-Seltzer™ Tablets

Catalog No. 19090788 Shop All Medique Products
Change view
Click to view available options
Quantity:
2/Pk., 36/Box
2/Pk.,18/Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-788 2/Pk., 36/Box
19-090-789 2/Pk.,18/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-090-788 Supplier Medique Supplier No. 13524
Add to Cart
Add to Cart

Effervescent pain reliever and antacid with specially buffered aspirin.

  • Antacid and pain reliever
  • Each Tablet contains Aspirin 324mg; Heat Treated Sodium Bicarbonate 1,916mg; Citric Acid (in water) 1,000mg

Specifications

Active Ingredients Aspirin (324 mg), Heat-treated Sodium Bicarbonate (1916 mg), Citric Acid (in water) (1000 mg)
For Use With (Application) Antacid and pain reliever
Product Type Buffered Aspirin Tablet
Format Tablet
Quantity 2/Pk., 36/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.