Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mediator of cell motility 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323408
Description
mediator of cell motility 1 Polyclonal antibody specifically detects mediator of cell motility 1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
mediator of cell motility 1 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 | |
C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1 | |
Mediator of cell motility 1 synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human mediator of cell motility 1. This peptide sequence is highly conserved in rat, mouse, and chicken Mmediator of cell motility 1. [UniProt# Q9Y316] | |
0.1 mL | |
Signal Transduction | |
51072 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/mL BSA and 50% Glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction