Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medegen Biohazardous Waste Bags, HDPE Film, Flat Pack
Shop All American Precision Plastics Products

Click to view available options
Dimensions (L x W):
30 x 43 in. (76.2 x 109.22cm)
33 x 40 in. (83.82 x 101.6cm)
40 x 48 in.
Thickness (Metric):
14 micron
16 micron
Description
- Made from HDPE
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with “Biohazardous Waste/Infectious Waste” and biohazard symbol imprinted in black ink
- Individually folded
Specifications
Specifications
| Capacity (English) | 40 to 45 gal. |
| Certifications/Compliance | ASTM Dart |
| Color | Red, Black |
| For Use With (Application) | Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab |
| Includes | Biohazardous or infectious waste with biohazard symbol |
| Material | HDPE |
| Thickness (Metric) | 16 micron |
| Dimensions (L x W) | 40 x 48 in. |
| Length (English) | 40 in. |
| Length (Metric) | 101.6 cm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction