Learn More
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll

Shop All American Precision Plastics Products


Description
- Made from low-density polyethylene
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
- Individually folded
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (Metric) | 45.4 to 60.6 L |
| Certifications/Compliance | ASTM Dart |
| Color | Red, Black |
| For Use With (Application) | Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab |
| Includes | Biohazardous or infectious waste with biohazard symbol |
| Material | LLDPE |
| Thickness (Metric) | 0.0304 mm |
| Thickness (English) | 1.2 mil |
| Dimensions (L x W) | 25 x 34 in. |
| Length (English) | 25 in. |
| Show More |
May NOT be used in CA, FL, PA or for interstate (between state) transport.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.