Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medegen Saf-T Tainer™ Biomedical Waste Container
SDP

Catalog No. 22278117 Shop All American Precision Plastics Products
Click to view available options
Capacity (English):
2.5 gal.
5 gal.
11 gal.
Diameter (English) Outer:
6 in.
12 in.
Height (English):
10.5 in.
12 in.
24 in.

Catalog No. 22-278117

Supplier: Medegen 102011

Only null left
Add to Cart
Add to Cart

A plastic liner sandwiched between corrugated walls makes this Saf-T Tainer™ Biomedical Waste Container leak and puncture resistant.

  • Rectangular, easy-to-assemble container stacks for convenient storage until incinerated
  • Integral handles for safe handling
  • Unassembled containers store flat to conserve space

Specifications

Capacity (Metric) 41.6 L
Design Infectious Waste with Biohazard Symbol
Diameter (Metric) Outer 30.48 cm
Diameter (English) Outer 12 in.
Capacity (English) 11 gal.
Color Red, Black
Height (Metric) 60.9 cm
Width (Metric) 30.4 cm
Liner Material Plastic
Width (English) 12 in.
Height (English) 24 in.
Product Type Biomedical Waste Container
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.