Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medline™ Multi-layer Lab Jackets

Catalog No. 19357711 Shop All Medline Industries Products
Click to view available options
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Protective and comfortable

  • Made with multi-layer nonwoven material that is fluid-resistant yet breathable
  • Features knit collar and cuffs, five snap-front closure, and three reinforced pockets
  • Falls just below the waist, proving less constrictive than lab coats
  • Unisex
  • Latex-free
  • White

Specifications

Size Large
Material SMS Fabric
Color Blue
Closure Type Snap
Construction Latex-free lab jackets, made with protective multilayer non-woven material
Collar Type Knit
Cuff Style Knit
Latex Free Yes
No. of Pockets 3
For Use With (Application) Lab purpose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.